Learn More
Troemner™ Individual Analytical Precision Weights, Class 2 with NVLAP Certificate
Designed for your most demanding weighing applications
Supplier:  Troemner™ 80858666
 View more versions of this product
Catalog No. 02-217-4BC
Only null left
Explore available promotions
Description
Description
Weights are designed so the specifications on surface finish, markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances decrease in order to enable the weight to maintain its tolerance over time.
  • Precision Analytical Individual Weights are made from the finest materials and are manufactured in sizes ranging from 1mg through 50kg
  • Fractional weights 5mg to 500mg are made of stainless steel (30mg weights are aluminum)
  • Fractional weights 3mg and below are made of aluminum
  • Weights are supplied in a rugged, precision-fit, high quality polycarbonate case
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
Metric Analytical Sheet Weight with One Side Turned Up
ASTM Class 2
2 mg
2 mg
Polycarbonate case
1
NVLAP, ISO9001
Aluminum
2.7g/cm3 at 20°C
Flat with one end turned up for easy handling
Each
Product Suggestions
Product Suggestions