Learn More
Troemner™ Individual Analytical Precision Weights, Class 2 with NVLAP Certificate
Designed for your most demanding weighing applications
$352.47 - $4607.59
Specifications
| Certifications/Compliance | NVLAP, ISO9001 |
|---|---|
| Classification | ASTM Class 2 |
| Includes | Polycarbonate case |
| Quantity | Each |
| Catalog Number | Mfr. No. | Mass | Density | Material | Shape | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Mass | Density | Material | Shape | Price | Quantity | ||
|
022172DC
|
Troemner™
80858242 |
10 g | 7.95g/cm3 at 20°C | Stainless Steel | Cylindrical with Handling Knob |
Each for $462.91
|
|
||
|
022173FC
|
Troemner™
80858331 |
500 mg | 7.95g/cm3 at 20°C | Stainless Steel | Flat with one end turned up for easy handling |
Each for $352.47
|
|
||
|
022173DC
|
Troemner™
80858445 |
100 mg | 7.95g/cm3 at 20°C | Stainless Steel | Flat with one end turned up for easy handling |
Each for $369.97
|
|
||
|
022173AC
|
Troemner™
80858574 |
10 mg | 7.95g/cm3 at 20°C | Stainless Steel | Flat with one end turned up for easy handling |
Each for $352.47
|
|
||
|
02-217-4BC
|
Troemner™
80858666 |
2 mg | 2.7g/cm3 at 20°C | Aluminum | Flat with one end turned up for easy handling |
Each for $352.47
|
|
||
|
02-214-119
|
Troemner™
80858371 |
300 mg | 7.95g/cm3 at 20°C | Stainless Steel | Flat with one end turned up for easy handling |
Each for $352.47
|
|
||
|
02-214-124
|
Troemner™
80858086 |
5 kg | 7.95g/cm3 at 20°C | Stainless Steel | Cylindrical with Handling Knob |
Each for $4,607.59
|
|
||
|
02-214-123
|
Troemner™
80858113 |
2 kg | 7.95g/cm3 at 20°C | Stainless Steel | Cylindrical with Handling Knob |
Each for $3,028.79
|
|
||
|
02-214-121
|
Troemner™
80858140 |
500 g | 7.95g/cm3 at 20°C | Stainless Steel | Cylindrical with Handling Knob |
Each for $1,067.59
|
|
||
|
02-214-122
|
N/A | 1 kg | 7.95g/cm3 at 20°C | Stainless Steel | Cylindrical with Handling Knob |
Each for $2,427.93
|
|
||
|
02-225-502
|
N/A | 50 g | 7.95g/cm3 at 20°C | Stainless Steel | Cylindrical with Handling Knob |
Each for $584.79
|
|
||
|
02-225-501
|
N/A | 100 g | 7.95g/cm3 at 20°C | Stainless Steel | Cylindrical with Handling Knob |
Each for $598.97
|
|
||
|
02-225-508
|
N/A | 2 g | 7.95g/cm3 at 20°C | Stainless Steel | Cylindrical with Handling Knob |
Each for $511.28
|
|
||
|
02-225-525
|
N/A | 5 mg | 7.95g/cm3 at 20°C | Stainless Steel | Flat with one end turned up for easy handling |
Each for $437.79
|
|
||
|
02-225-548
|
N/A | 200 mg | 7.95g/cm3 at 20°C | Stainless Steel | Flat with one end turned up for easy handling |
Each for $437.79
|
|
||
|
02-225-549
|
N/A | 1 g | 7.95g/cm3 at 20°C | Stainless Steel | Cylindrical with Handling Knob |
Each for $477.52
|
|
||
Description
Weights are designed so the specifications on surface finish, markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances decrease in order to enable the weight to maintain its tolerance over time.
- Precision Analytical Individual Weights are made from the finest materials and are manufactured in sizes ranging from 1mg through 50kg
- Fractional weights 5mg to 500mg are made of stainless steel (30mg weights are aluminum)
- Fractional weights 3mg and below are made of aluminum
- Weights are supplied in a rugged, precision-fit, high quality polycarbonate case
Specifications
| NVLAP, ISO9001 | |
| ASTM Class 2 | |
| Polycarbonate case | |
| Each |