Learn More
Thermo Scientific Chemicals TRIS, 1.0M buffer soln., pH 7.5, 0.2 micron filtered
CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
Supplier: Thermo Scientific Chemicals J62993AP
Description
Tris Buffered Saline (TBS) is a pH stabilizing solution used for western blot and ELISA procedures. It is used as a convenient prepared buffer for use in various applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTris Buffered Saline (TBS) is a pH stabilizing solution used for western blot and ELISA procedures. It is used as a convenient prepared buffer for use in various applications.
Solubility
It is soluble in water.
- Environmental benefits include:
- Renewable Energy
Specifications
TRIS | |
77-86-1 | |
7.5 | |
500 mL | |
Odorless | |
It is soluble in water. | |
C(C(CO)(CO)N)O | |
121.136 | |
CHEBI:9754 |
0.2 micron filtered | |
1.0 M | |
Liquid | |
C4H11NO3 | |
MFCD00004679 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol | |
6503 |
Chemical Identifiers
77-86-1 | |
121.136 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
CHEBI:9754 | |
C(C(CO)(CO)N)O |
C4H11NO3 | |
MFCD00004679 | |
6503 | |
2-amino-2-(hydroxymethyl)propane-1,3-diol |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only