missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin ICP Standard, 10,000 ppm Sn in 20% HCl, Ricca Chemical
Chemical Identifiers
CAS | 7440-31-5 |
---|---|
Molecular Formula | Sn |
Molecular Weight (g/mol) | 118.71 |
MDL Number | MFCD00133862 |
InChI Key | ATJFFYVFTNAWJD-UHFFFAOYSA-N |
PubChem CID | 5352426 |
ChEBI | CHEBI:32990 |
IUPAC Name | tin |
SMILES | [Sn] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
PSN10KH-100
|
Ricca Chemical Company
PSN10KH100 |
100 mL | Natural LDPE Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
PSN10KH-500
|
Ricca Chemical Company
PSN10KH500 |
500 mL | Natural Poly Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
7440-31-5 | |
118.71 | |
ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
CHEBI:32990 | |
[Sn] |
Sn | |
MFCD00133862 | |
5352426 | |
tin |
Specifications
Natural LDPE Bottle | |
7440-31-5,7647-01-0,7732-18-5 | |
10,000ppm Sn | |
1 mL = 10 mg Sn | |
MFCD00133862 | |
[Sn] | |
118.71 | |
CHEBI:32990 |
100 mL | |
77.0,19.65,0.78 | |
81.0,20.45,0.81 | |
Sn | |
ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
tin | |
5352426 | |
Tin ICP Standard |