missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin ICP Standard, 10,000 ppm Sn in 20% HCl, Ricca Chemical
Supplier: Ricca Chemical Company PSN10KH100
Specifications
| Tin ICP Standard | |
| 7440-31-5,7647-01-0,7732-18-5 | |
| 10,000ppm Sn | |
| 1 mL = 10 mg Sn | |
| MFCD00133862 | |
| [Sn] | |
| 118.71 | |
| CHEBI:32990 |
| Natural LDPE Bottle | |
| 77.0,19.65,0.78 | |
| 81.0,20.45,0.81 | |
| Sn | |
| ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
| tin | |
| 5352426 |
Chemical Identifiers
| 7440-31-5 | |
| 118.71 | |
| ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
| CHEBI:32990 | |
| [Sn] |
| Sn | |
| MFCD00133862 | |
| 5352426 | |
| tin |