missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin ICP Standard, 10,000 ppm Sn in 20% HCl, Ricca Chemical
Supplier: Ricca Chemical Company PSN10KH500
Description
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Tin ICP Standard | |
500 mL | |
77.0,19.65,0.78 | |
81.0,20.45,0.81 | |
Sn | |
ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
tin | |
5352426 |
Natural Poly Bottle | |
7440-31-5,7647-01-0,7732-18-5 | |
10,000ppm Sn | |
1 mL = 10 mg Sn | |
MFCD00133862 | |
[Sn] | |
118.71 | |
CHEBI:32990 |
Chemical Identifiers
7440-31-5 | |
118.71 | |
ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
CHEBI:32990 | |
[Sn] |
Sn | |
MFCD00133862 | |
5352426 | |
tin |