missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tetra Core Set and Bolt™ Bis-Tris Plus 4-12% Gels Combos
GREENER_CHOICE

The Tetra Electrophoresis Core Set and Bolt Bis-Tris Plus 4-12% Combo includes the Tetra Electrophoresis primary and companion cores along with three boxes of Bolt Bis-Tris Plus gels. Easily swap and run up to four Bolt Bis-Tris gels in Bio-Rad's Mini-PROTEAN™ Tetra Cell tank with a run time as short as 20 minutes.

$758.00 - $758.00

Specifications

Adapters Tetra Electrophoresis Cores
Content And Storage Store at 4°C to 25°C. Do not freeze.
Buffer Bolt MES SDS Running Buffer, Bolt MOPS SDS Running Buffer
For Use With (Equipment) Bio-Rad Mini Protean Tanks
Gel Percentage 4 to 12%
View More Specs

Products 3
Catalog Number Mfr. No. Wells Price Quantity  
Catalog Number Mfr. No. Wells Price Quantity  
NW0412TS10W
View Documents
Invitrogen™
NW0412TS10W
10-well
Each for $758.00
Only null left
 
NW0412TS12W
View Documents
Invitrogen™
NW0412TS12W
12-well
Each for $758.00
Only null left
 
NW0412TS15W
View Documents
Invitrogen™
NW0412TS15W
15-well
Each for $758.00
Only null left
 
Description
Show More
Specifications

Specifications

Tetra Electrophoresis Cores
Bolt MES SDS Running Buffer, Bolt MOPS SDS Running Buffer
4 to 12%
1.0 mm
Electrophoresis Cores and Precast Protein Gels
3.5 to 260 kDa
Molecular Weight
Tetra Core Set, 3 boxes of gels (30 gels)
Store at 4°C to 25°C. Do not freeze.
Bio-Rad Mini Protean Tanks
Mini
Bis-Tris Plus
Protein
Denaturing
Bolt™, WedgeWell™
8 cm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only.