missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tantalum Standard For ICP, 1000μg/mL in H2O/0.8% HF, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Manufacturer:  Spex Certiprep Llc PLTA92X

 View more versions of this product

Catalog No. PLTA9-2X



Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ta1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Tantalum Standard
Ta
[Ta]
180.948
CHEBI:33348
Triple-leached LDPE bottle
500mL
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
7440-25-7,7664-39-3,7732-18-5
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum
23956
Assurance
H2O/0.8% HF
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Chemical Identifiers

7440-25-7
180.948
23956
tantalum
Ta
GUVRBAGPIYLISA-UHFFFAOYSA-N
CHEBI:33348
[Ta]
Safety and Handling

Safety and Handling

Toxic in contact with skin. Causes serious eye irritation

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents
Product Certifications
Promotions

Promotions

Product intended for laboratory use only