missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tantalum Standard For ICP, 1000μg/mL in H2O/0.8% HF, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$277.44 - $458.85

Chemical Identifiers

CAS 7440-25-7
Molecular Formula Ta
Molecular Weight (g/mol) 180.95
MDL Number MFCD00011252
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
PubChem CID 23956
ChEBI CHEBI:33348
IUPAC Name tantalum
SMILES [Ta]
View More Specs

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PLTA9-2Y
View Documents Promotion Details
SPEX CertiPrep Group LLC
PLTA92Y
125 mL Triple-leached LDPE bottle N/A
 
PLTA9-2X
View Documents Promotion Details
SPEX CertiPrep Group LLC
PLTA92X
500 mL Triple-leached LDPE bottle N/A
 
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ta1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

7440-25-7
180.95
GUVRBAGPIYLISA-UHFFFAOYSA-N
CHEBI:33348
[Ta]
Ta
MFCD00011252
23956
tantalum

Specifications

7440-25-7,7664-39-3,7732-18-5
MFCD00011252
[Ta]
180.95
CHEBI:33348
Triple-leached LDPE bottle
125 mL
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Ta
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum
23956
Assurance
H2O/0.8% HF
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Tantalum Standard
Safety and Handling

Safety and Handling

Toxic in contact with skin. Causes serious eye irritation

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
missing translation for 'documents'

missing translation for 'documents'

Product intended for laboratory use only