Learn More
Cole-Parmerâ„¢ Vial Holder for HG-600 Geno/Grinderâ„¢ 2010 Homogenizer
This Vial Holder for HG-600 Geno/Grinderâ„¢ 2010 Homogenizer securely hold vials in place while processing samples.
$175.31 - $298.03
Specifications
Product Type Holder
For Use With (Equipment) HG-600 Geno/Grinderâ„¢ 2010 Homogenizer
Material Foam
Packaging Quantity 1 Ea.
View More Specs
Products 2
Catalog Number Mfr. No. Length (Metric) Holds Price Quantity  
Catalog Number Mfr. No. Length (Metric) Holds Price Quantity  
2197
View Documents
Cole-Parmerâ„¢
SPX2197
11.9 cm 24 Standard 15 mL Conical Bottom Centrifuge Tubes
Each for $175.31
Only null left
2196
View Documents
Cole-Parmerâ„¢
219616PE
11.5 cm 12 Standard 50 mL Conical Bottom Centrifuge Tubes
Each for $298.03
Only null left
Description
Description
  • Safely grind samples in the HG-600 Geno/Grinderâ„¢ 2010.
  • Vial holders are suitable for use with the HG-600 Geno/Grinderâ„¢ 2010 Homogenizer.
  • Order homogenizer and vials separately.
Specifications
Specifications
Holder
HG-600 Geno/Grinderâ„¢ 2010 Homogenizer
Foam
1 Ea.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'