Learn More
MilliporeSigma™ Supelco™ SGE Needles for Luer Lock Syringes
Preferred for manual injection where piercing the septum in exactly the same place is difficult. The beveled tip is designed for optimum septum penetration and prevention of septum coring.
Supplier: MilliporeSigma™ Supelco™ 26271
Description
Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
Stainless Steel (Tip) | |
23 G | |
SGE Needles | |
2 Pk. | |
Cone with Side Port |
NLL-5/23 H | |
0.63 mm | |
5 | |
50 mm | |
Luer Lock |