Learn More
Selenium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$248.29 - $496.59
Chemical Identifiers
CAS | 7782-49-2 |
---|---|
Molecular Formula | Se |
Molecular Weight (g/mol) | 78.97 |
MDL Number | MFCD00134090 MFCD00011224 |
InChI Key | BUGBHKTXTAQXES-UHFFFAOYSA-N |
PubChem CID | 6326970 |
ChEBI | CHEBI:27568 |
IUPAC Name | selenium |
SMILES | [Se] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
PLSE2-2Y
|
SPEX CertiPrep Group LLC
PLSE22Y |
125 mL | Triple-leached LDPE bottle |
Each for $248.29
|
|
|||||
PLSE2-2X
|
SPEX CertiPrep Group LLC
PLSE22X |
500 mL | Triple-leached LDPE bottle |
Each for $496.59
|
|
|||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Se | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7782-49-2 | |
78.97 | |
BUGBHKTXTAQXES-UHFFFAOYSA-N | |
CHEBI:27568 | |
[Se] |
Se | |
MFCD00134090 MFCD00011224 | |
6326970 | |
selenium |
Specifications
Triple-leached LDPE bottle | |
Assurance Certificate of Analysis reports up to 30 elements at ppb levels. | |
1.0,5.0,94.0 | |
Se | |
AA, ICP | |
Certified Reference Material | |
[Se] | |
selenium | |
125 mL | |
CHEBI:27568 | |
Matrix Concentration Cat. No. 2-5% HNO3 1000μg/mL PLSE2-2Y, -2T, -2X 10,000 PLSE2-3Y, -3X |
7782-49-2,7697-37-2,7732-18-5 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
1.0,5.0,94.0 | |
MFCD00134090 MFCD00011224 | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
BUGBHKTXTAQXES-UHFFFAOYSA-N | |
2% HNO3 | |
78.97 | |
6326970 | |
Assurance | |
Assurance Grade Selenium Standard |
Safety and Handling
Danger: Causes severe skin burns and eye damage Do not breathe dusts or mists. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only