missing translation for 'onlineSavingsMsg'
Learn More

Selenium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLSE22X

 View more versions of this product

Catalog No. PLSE2-2X

Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Se1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Assurance Grade Selenium Standard
7782-49-2,7697-37-2,7732-18-5
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
1.0,5.0,94.0
MFCD00134090 MFCD00011224
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
BUGBHKTXTAQXES-UHFFFAOYSA-N
2% HNO3
78.97
6326970
Assurance
Triple-leached LDPE bottle
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
1.0,5.0,94.0
Se
AA, ICP
Certified Reference Material
[Se]
selenium
500 mL
CHEBI:27568
Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLSE2-2Y, -2T, -2X    10,000    PLSE2-3Y, -3X

Chemical Identifiers

7782-49-2
78.97
BUGBHKTXTAQXES-UHFFFAOYSA-N
CHEBI:27568
[Se]
Se
MFCD00134090 MFCD00011224
6326970
selenium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Danger: Causes severe skin burns and eye damage Do not breathe dusts or mists. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only