missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Interface Module for Cubis Balances
GREENER_CHOICE

Interface Module for use with Cubis balances.

$409.02 - $417.37

Specifications

For Use With (Equipment) Cubis Balance
Type Data Interface
View More Specs

Products 3
Catalog Number Mfr. No. Connection Type For Use With (Application) Price Quantity  
Catalog Number Mfr. No. Connection Type For Use With (Application) Price Quantity  
14-557-550
GREENER_CHOICE
View Documents
Sartorius
YDO01MSP
Option P: RS-232 9 Pin, PS/2 Connecting a PC or a keyboard
Each for $417.37
Only null left
 
14-557-548
GREENER_CHOICE
View Documents
Sartorius
YDO01MSR
Option R: RS-232C Data Interface, 25 Pin Connection of Cubis accessory equipment
Each for $417.37
Only null left
 
14-557-549
View Documents
Sartorius
YDO01MSB
Option B: Bluetooth Adapter Wireless connection of the YDP10BT data printer
Each for $409.02
Only null left
 
Description

Description

  • Option R (YDO01MS-R): RS-232C data interface, 25-pin, for connection of Cubis accessory equipment
  • Option B (YDO01MS-B): Bluetooth™ interface module for wireless connection of the YDP10BT data printer
  • Option P (YDO01MS-P): RS-232C data interface, 9-pin, including PS/2 port for connecting a PC or a keyboard
Specifications

Specifications

Cubis Balance
Data Interface
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'