Learn More
Sartorius™ Cellulose Acetate Membrane Filters
Combines high flow rates and thermal stability with very low adsorption characteristics. Sartorius™ Cellulose Acetate Membrane Filters are excellently suited for use in pressure filtration devices. Non-sterile filters in a variety of diameters and pore sizes are bulk-packed to save money and space.
$34.00 - $550.00
Specifications
Material | Cellulose Acetate |
---|---|
For Use With (Application) | For use in pressure filtration devices; Sterile filtration of solution. |
Thickness | 120µm |
Color | White |
Sterility | Non-sterile |
Catalog Number | Mfr. No. | Pore Size | Diameter (Metric) Outer | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Pore Size | Diameter (Metric) Outer | Price | Quantity | ||||
14-555-352
|
Sartorius™
1110447N |
0.8μm | 47mm |
Pack of 100 Each for $117.00
|
|
14-555-353
|
Sartorius™
1110450N |
0.8μm | 50mm |
Pack of 100 Each for $131.00
|
|
14-555-351
|
Sartorius™
1110425N |
0.8μm | 25mm |
Pack of 100 Each for $255.00
|
|
14-555-354
|
Sartorius™
1110490N |
0.8μm | 90mm |
Pack of 100 Each for $93.00
|
|
14-555-356
|
Sartorius™
1110547N |
0.65μm | 47mm |
Pack of 100 Each for $83.00
|
|
14-555-357
|
Sartorius™
1110550N |
0.65μm | 50mm |
Pack of 100 Each for $83.00
|
|
14-555-358
|
Sartorius™
11106100G |
0.45μm | 100mm |
Pack of 25 Each for $39.00
|
|
14-555-361
|
Sartorius™
1110630N |
0.45μm | 30mm |
Pack of 100 Each for $34.00
|
|
14-555-363
|
Sartorius™
1110650N |
0.45μm | 50mm |
Pack of 100 Each for $74.00
|
|
14-555-359
|
Sartorius™
1110613N |
0.45μm | 13mm |
Pack of 100 Each for $39.00
|
|
14-555-360
|
Sartorius™
1110625N |
0.45μm | 25mm |
Pack of 100 Each for $39.00
|
|
14-555-365
|
Sartorius™
1110690G |
0.45μm | 90mm |
Pack of 25 Each for $144.00
|
|
14-555-364
|
Sartorius™
1110685N |
0.45μm | 85mm |
Pack of 100 Each for $550.00
|
|
14-555-362
|
Sartorius™
1110647N |
0.45μm | 47mm |
Pack of 100 Each for $123.00
|
|
14-555-366
|
Sartorius™
1110713N |
0.2μm | 13mm |
Pack of 100 Each for $84.00
|
|
14-555-367
|
Sartorius™
1110725N |
0.2μm | 25mm |
Pack of 100 Each for $96.00
|
|
14-555-368
|
Sartorius™
1110730N |
0.2μm | 30mm |
Pack of 100 Each for $128.00
|
|
14-555-369
|
Sartorius™
1110747N |
0.2μm | 47mm |
Pack of 100 Each for $125.00
|
|
14-555-370
|
Sartorius™
1110750N |
0.2μm | 50mm |
Pack of 100 Each for $125.00
|
|
14-555-371
|
Sartorius™
1110790G |
0.2μm | 90mm |
Pack of 25 Each for $144.00
|
|
14-555-431
|
Sartorius™
12303100G |
1.2μm | 100mm |
Pack of 25 Each for $425.00
|
|
14-555-432
|
Sartorius™
1230325N |
1.2μm | 25mm |
Pack of 100 Each for $425.00
|
|
14-555-433
|
Sartorius™
1230347N |
1.2μm | 47mm |
Pack of 100 Each for $425.00
|
|
14-555-435
|
Sartorius™
1234247N |
5.0μm | 47mm |
Pack of 100 Each for $220.00
|
|
14-555-434
|
Sartorius™
1234225N |
5.0μm | 25mm |
Pack of 100 Each for $161.00
|
|
14-555-355
|
Sartorius™
1110525N |
0.65μm | 25mm |
Pack of 100 Each for $83.00
|
|
Description
Combines high flow rates and thermal stability with very low adsorption characteristics. Sartorius™ Cellulose Acetate Membrane Filters are excellently suited for use in pressure filtration devices. Non-sterile filters in a variety of diameters and pore sizes are bulk-packed to save money and space.
- Ideal for colony counting, particle testing, microscopy and sterility testing
- High-quality CA membrane filters for use in pressure filtration devices
- Available in pore sizes ranging from 0.2μm up to 5.0μm
- 0.2μm filters are the standard for sterile filtration of aqueous solutions, such as nutrient media, buffers and sera
- 5μm filters can be used as prefilters to retain coarse particles, while allowing microorganisms through
- White, non-sterile filters without grids in a variety of diameters (13 to 100mm)
- Can be sterilized by autoclaving at 121°C or 134°C; gamma irradiated to 25 kGy, dry heat or ethvlene oxide
The results of publications on adsorption are difficult to correlate, as mostly different test substances, conditions and detection methods were used, and the membranes were tested without previously being sterilized.
Specifications
Cellulose Acetate | |
120µm | |
Non-sterile |
For use in pressure filtration devices; Sterile filtration of solution. | |
White | |
Yes |