missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Sartolab™ P20 Plus Filter Systems

Filter tissue culture solutions quickly, economically and reliably. Sartorius Sartolab™ P20 Plus Filter Systems are sterile, ready-to-use units. A large filtration area and automatic de-aeration ensures high flow rates. Available with PES or SFCA membrane, different connectors or with a prefilter to process up to 10L.

Supplier:  Sartorius 18075D

 View more versions of this product

Catalog No. 14-555-991

Only null left
Explore available promotions

Description

Description

Filter tissue culture solutions quickly, economically and reliably. Sartorius Sartolab™ P20 Plus Filter Systems are sterile, ready-to-use units. A large filtration area and automatic de-aeration ensures high flow rates. Available with PES or SFCA membrane, different connectors or with a prefilter to process up to 10L.

The new way for reliable sterile filtration
  • Ready-to-use unit with a large filtration area (20cm2) for fast filtration
  • Quickly and reliably sterilizes media and aqueous solutions with optimal throughputs
  • Automatically vents any trapped air through the PTFE membrane-protected vent ports
  • Extraordinarily favorable price-performance ratio for a cost-effective solution
  • The total filterable volume can often be doubled due to an integrated binder-free, glass fiber pre-filter
  • High-quality construction; every batch is rigorously tested to ensure sterility and integrity
Specifications

Specifications

USP, Class VI
Single-use
Polycarbonate
0.2 μm
Sterile
Reliable sterile filtration of tissue culture solutions
3 bar
10 Pack
100 mL to 5 L
Luer Lock, Hose Nipple
Ready-to-Use Pressure Filter
PES
100 mL to 5 L
20 cm2
Glass Prefilter, Filling Bell
5 bar
Pressure Filter
0.3 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions