Learn More
Corning™ PYREX™ HPLC Reservoir, Single Cavity
Reservoirs have a central cavity for a mobile phase inlet reservoir
Supplier:  Corning™ 21502L
 View more versions of this product
Catalog No. 13-645-096
Only null left
Explore available promotions
Description
Description
Reservoirs have a central cavity for a mobile phase inlet reservoir
PYREX HPLC reservoirs have a central cavity for a mobile phase inlet reservoir, an autoclavable plastic coat that absorbs UV up to 385 nm, and a GL 45 thread finish. These may be used with our 3-hole cap (Cat. No. 1395-3H).
Order Info
Optional 3-hole cap available separately (13-645-099)
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Specifications
Specifications
5.9 in.
Plastic
Reservoirs have a central cavity for a mobile phase inlet reservoir
325 mm
1/Pk.
150 mm
2 L
HPLC Reservoir
12.7 in.
1/Cs.
Autoclavable
Product Suggestions
Product Suggestions