Learn More
Promega Restriction Enzymes - Sal I Blue/White Cloning and Genome Qualified (D)
Capable of digesting DNA in 15 minutes or less. Genome qualified to ensure optimal performance in genomic analysis applications. (G/TCGA C C AGCT\G)
Supplier: Promega R6051
Description
- Capable of digesting DNA in 15 minutes or less
- Active and capable of digestion directly in GoTaq(R) Green Master Mix
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
- Genome qualified to ensure optimal performance in genomic analysis applications
Specifications
10 U/μL | |
37°C | |
Provides a higher level of quality control for Enzymes used in cloning applications | |
-30°C to -10°C | |
G.TCGAC CAGCT.G | |
Liquid |
1 x 1mL 10X Buffer D, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 2000U Sall | |
7.4 | |
10mM Tris HCl (pH 7.4), 300mM KCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol | |
2000 U | |
SalI |