missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Restriction Enzymes - Sal I Blue/White Cloning and Genome Qualified (D)
Capable of digesting DNA in 15 minutes or less. Genome qualified to ensure optimal performance in genomic analysis applications. (G/TCGA C C AGCT\G)
$98.96 - $364.47
Specifications
| Incubator Temperature | 37°C |
|---|---|
| For Use With (Application) | Provides a higher level of quality control for Enzymes used in cloning applications |
| Storage Buffer | 10mM Tris HCl (pH 7.4), 300mM KCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol |
| Cut Site | G.TCGAC CAGCT.G |
| Form | Liquid |
Description
- Capable of digesting DNA in 15 minutes or less
- Active and capable of digestion directly in GoTaq(R) Green Master Mix
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
- Genome qualified to ensure optimal performance in genomic analysis applications
‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.
Specifications
| 37°C | |
| 10mM Tris HCl (pH 7.4), 300mM KCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol | |
| Liquid |
| Provides a higher level of quality control for Enzymes used in cloning applications | |
| G.TCGAC CAGCT.G |