Learn More
Promega HaloTag™ Mammalian Pull-Down Systems
The HaloTag™ Mammalian Pull-Down Systems capture and purify intracellular protein complexes for correlative cellular localization and real-time imaging studies with the same genetic construct for further understanding of overall protein function.
Supplier: Promega G6500
Includes: Buffers and resin necessary to perform a HaloTag pull-down and HaloTag TMRDirect™ Ligand
Description
- Identify more Physiologically Relevant Protein Partners
- Rapid capture of simple and complex protein pairs directly from lysates
- Move from pull-downs to imaging studies using same construct
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
COMPONENTS AT DIFFERENT TEMPS | |
HaloTag Mammalian Pull-down System | |
24 Reactions | |
Buffers and Resin necessary to perform a HaloTag™ pull-down and HaloTag™ TMRDirect™ ligand |