Learn More
Promega Flexi™ Enzyme Blends DFS Item
The 10X Flexi Enzyme Blend (SgfI and PmeI) is required to screen Flexi Vector clones for the presence of the protein-coding region of interest. This enzyme blend can also be used to cleave PCR products prior to cloning into the Flexi Vectors or transfer p
Supplier:  Promega R1851
 View more versions of this product
Catalog No. PR-R1851
Only null left
Explore available promotions
Description
Description
  • Reagent Used with Flexi(R) Cloning System
  • Required to screen Flexi(R) Vector clones for presence of protein-coding region of interest
  • Also can be used to cleave PCR products prior to cloning into Flexi(R) Vectors or transfer protein-coding regions between Flexi(R) Vectors
Specifications
Specifications
-30°C to -10°C
10X Flexi Enzyme blend (SgfI and Pmel)1 x 100μL, Nuclease-free Water 2 x 1,250μL, 5X Flexi Digest Buffer 1 x 1mL
Enzyme Blend
Fluid
Flexi cloning system
7.5
25 μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Licensed under U.S. Patent 5,945,288.