missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Phosphine-PEG3-Biotin

Catalog No. PI88901
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88901 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI88901 Supplier Thermo Scientific™ Supplier No. 88901

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Label azide-tagged molecules for metabolic labeling (Staudinger ligation) and streptavidin detection using this phosphine-activated biotinylation reagent.

Thermo Scientific™ Pierce Biotin-PEG3-Phosphine is a biotinylation reagent for labeling azide-containing molecules, which enables biotin-based detection and affinity purification of molecules via Staudinger ligation strategies.

The phosphine group reacts with an azide to produce an aza-ylide intermediate that is trapped to form a stable, covalent amide bond, which is also referred to as the Staudinger reaction. The azide group, which is the reactive target of phosphine-activated biotin, can be introduced into proteins or other cellular targets via azide reagents or through in vivo labeling with azide-derivatives of naturally occurring metabolic building blocks. Because neither phosphines nor azides are present in biological systems, they comprise chemoselective (mutually specific) ligation pairs for labeling and conjugation. Thus, when used in combination with azide labeling strategies, Phosphine-PEG3-Biotin enables directed biotin labeling to facilitate detection or affinity purification of protein interactions and post-translational modifications using streptavidin probes or streptavidin agarose resins.

Highlights:

  • Soluble easily dissolves in water-miscible solvents (e.g., DMSO) for subsequent dilution in aqueous reaction mixtures with cell lysates and other biological samples

  • Compatible reaction chemistry occurs effectively in simple buffer conditions; requires no accessory reagents such as copper or reducing agents

  • Chemoselective the phosphine reactive group is specific in biological samples for azide-tagged molecules, ensuring that biotinylation is specific

  • PEG spacer polyethylene glycol spacer arm helps maintain solubility of labeled molecules and decreases steric hindrance for affinity-binding to avidin, streptavidin or NeutrAvidin Protein

Specifications

Product Type Phosphine-PEG3-Biotin
Quantity 10 mg
Content And Storage Store at -20°C. Shipped at ambient temperature.
Product Line EZ-Link
Cell Permeability Cell-Impermeant
Chemical Reactivity Azide
Reactive Moiety Phosphine
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye Biotin
Spacer Extra-long, Pegylated
Label Type Biotin & Analogs
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.