missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PDPK1 (Ser241) Recombinant Rabbit Monoclonal Antibody (PDK1S241-F7), PE, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
$380.36 - $935.64
Specifications
Antigen | Phospho-PDPK1 (Ser241) |
---|---|
Clone | PDK1S241-F7 |
Applications | Flow Cytometry |
Classification | Recombinant Monoclonal |
Conjugate | PE |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
PIMA537338
|
Thermo Fisher Scientific
MA537338 |
10 Tests |
Each for $380.36
|
|
||||||
PIMA537089
|
Thermo Fisher Scientific
MA537089 |
100 Tests |
Each for $935.64
|
|
||||||
Description
PDPK1 (3 Phosphoinositide Dependent Protein Kinase 1) phosphorylates AGC kinases. PDPK1 activates conventional PKC and PKC zeta through phosphorylation of critical threonine residues in the activation loop. PDPK1 also phosphorylates Protein Kinase B (PKB) at threonine 308 in the presence of phosphatidylinositol-3,4,5-trisphosphate. Active Akt inactivates Glycogen Synthase Kinase 3 (GSK3), eventually leading to the dephosphorylation and activation of glycogen synthase, and the stimulation of glycogen synthesis. Because of the role that PDPK1 plays in insulin-induced glycogen synthesis and PKC activation, it is a potentially important target for metabolic drug research.Specifications
Phospho-PDPK1 (Ser241) | |
Flow Cytometry | |
PE | |
Rabbit | |
Human | |
O15530 | |
5170 | |
A synthetic phospho-peptide corresponding to residues surrounding Ser241 of human phospho PDPK1. | |
Primary | |
4°C | |
Pdpk1 |
PDK1S241-F7 | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
PBS with 0.20% BSA and 0.09% sodium azide | |
3-phosphoinositide dependent protein kinase 1; 3-phosphoinositide dependent protein kinase-1; 3-phosphoinositide-dependent protein kinase 1; 3-phosphoinositide-dependent protein kinase 2 pseudogene; HGNC:8809; hPDK1; isoenzyme 1; kinase isoenzyme 1; lipoamide; mPDK1; OTTHUMP00000174525; OTTHUMP00000205076; PDK1; PDPK1; PDPK2; PDPK2P; Pkb kinase; PkB kinase like gene 1; PkB-like 1; PRO0461; Protein kinase B kinase | |
Pdpk1 | |
IgG Îş | |
Protein A/G | |
Antibody |