missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBS, Phosphate Buffered Saline, 1X Solution, pH 7.4, Fisher BioReagents™

$241.08 - $737.00
Specifications
Content And Storage | Room temperature |
---|---|
Packaging Type | Poly Cube |
Melting Point | 0°C |
pH | 7.3 to 7.5 |
Form | Liquid |
Description
PBS, Phosphate Buffered Saline, 1X Solution, pH 7.4
A standard phosphate buffer used in many biomolecular procedures.Specifications
Room temperature | |
0°C | |
Liquid |
Poly Cube | |
7.3 to 7.5 |