Learn More
Thermo Scientific™ Nalgene™ Reusable Filter Units
GREENER_CHOICE
Eliminate the need for a manifold or breakable glass vacuum flask. Nalgene Reusable Filter Holders with Receiver are complete, environmentally-friendly systems.
Supplier:  Thermo Scientific™ 3004100
 View more versions of this product
Catalog No. 09-740-23E
Only null left
Explore available promotions
Description
Description
Reusable filter units include two membrane support plates: a clear one-piece plate for sterilization applications and a two-piece white plate for analytical applications. Accepts 47 or 50 mm diameter membranes. Two side-arms allow connection to vacuum line. Tubing adapters accept 1/4- to 5/16-inch (6 to 8 mm) I.D. vacuum tubing. 4 units per case.
Features
  • Replaces metal or breakable glass – major components are made of durable, transparent, autoclavable polysulfone
  • Use for sterilization or analysis. Filter holders come with sterilization and analytical membrane support plates
  • Designed to be used with vacuum or pressure
  • Graduated chambers for easy reference
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
Autoclavable
1
500 mL
Polysulfone
1000 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Schematics & Diagrams