missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Copolymer (PPCO) Lowboys with Spigot

Dispense most liquids, including reagents, distilled water and titrants, using these polypropylene copolymer (PPCO) lowboys, which fit into tight spaces and can stack three high.

Supplier:  Thermo Scientific™ 23240015

 View more versions of this product

Catalog No. 02-963-4C



Only null left
Explore available promotions

Alert:

Disengage threads and dismount spigot before autoclaving

Follow autoclave instructions provided with the product for proper function

Note: Tubing adapter is not autoclavable


Description

Description

Create a compact, space saving, reagent dispensing bank using chemical-resistant Thermo Scientific™ Nalgene™ Low-Profile autoclavable polypropylene copolymer (PPCO) Lowboys. The low-profile design of these high density polyethylene lowboys fits into tight spaces on shelves and in refrigerators and can stack three high. Chemically compatible with a wide range of lab liquids. Convenient integral carry handle for safe, easy transport. Nalgene polypropylene copolymer carboys can be autoclaved before refilling to eliminate bacterial growth when storing sterile solutions. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.

  • Polypropylene spigot and screw closure
  • Contour handle allows easy one-handed carrying — even when full
  • Can tolerate strong acids and bases for a wide variety of solutions
  • Follow autoclave instructions provided with the product to ensure proper function
  • Autoclavable/Leakproof†

Includes:
Polypropylene screw closures

Greener Choice Claims
  • Environmental benefits include:
    • Source Reduction
Specifications

Specifications

Carboy with Spigot
Autoclavable Lowboy w/Spigot PP 15 L
4 gal.
371 mm
44 mm
8.27 in.
4
63 mm
PPCO
15 L
1.73 in.
371 mm
210 mm
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Manuals

Promotions

Promotions

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.