missing translation for 'onlineSavingsMsg'
Learn More

Mineral Oil, Medium and Low Viscosity, FCC, Spectrum™ Chemical
GREENER_CHOICE

M1194, 8042-47-5

$301.03 - $3,141.69

Chemical Identifiers

CAS 8042-47-5
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18604622
View Documents
Spectrum Chemical Mfg Cor
M1194500MLGL
500 mL Amber Glass Bottle N/A
Only null left
 
18604618
View Documents
Spectrum Chemical Mfg Cor
M11941LT
1 L Amber Glass Bottle N/A
Only null left
 
18604619
View Documents
Spectrum Chemical Mfg Cor
M11944LTGL
4 L Amber Glass Bottle N/A
Only null left
 
18604620
View Documents
Spectrum Chemical Mfg Cor
M11944LTGLCS4
4 x 4 L Amber Glass Bottle N/A
Only null left
 
18604621
View Documents
Spectrum Chemical Mfg Cor
M119420LT
20 L Poly Drum N/A
Only null left
 
Description

Description

Spectrum™ Chemical Mineral Oil, Medium and Low Viscosity, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

8042-47-5

Specifications

8042-47-5
Amber Glass Bottle
0.848-0.867
FCC
1
500 mL
3-11 cSt
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'