Learn More
Mineral Oil, Medium and Low Viscosity, FCC, Spectrum™ Chemical
M1194, 8042-47-5
Supplier:  Spectrum Chemical Mfg Cor M11944LTGLCS4
 View more versions of this product
Catalog No. 18604620
Only null left
Explore available promotions
Description
Description
Spectrum™ Chemical Mineral Oil, Medium and Low Viscosity, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
8042-47-5
Amber Glass Bottle
0.848-0.867
1
4 x 4 L
3-11 cSt
Chemical Identifiers
8042-47-5
Product Suggestions
Product Suggestions