missing translation for 'onlineSavingsMsg'
Learn More

Mineral Oil, Medium and Low Viscosity, FCC, Spectrum™ Chemical

M1194, 8042-47-5

Supplier:  Spectrum Chemical Mfg Cor M11941LT

 View more versions of this product

Catalog No. 18604618


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Mineral Oil, Medium and Low Viscosity, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

8042-47-5
Amber Glass Bottle
0.848-0.867
FCC
1
1 L
3-11 cSt

Chemical Identifiers

8042-47-5
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions