missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 9mm Screwthread Autosampler Vials With Insert

MicroLiter 9mm autosampler screwthread vials with inserts are identical, clean, and dependable, and are certified to ensure optimal performance and reproducible results.

Supplier:  DWK Life Sciences 091200102

 View more versions of this product

Catalog No. 15-111-227


Only null left
Explore available promotions

Description

Description

Performance equal to an 11 mm crimp-top vial. Inserts available
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
    • Non-Toxic
    • Ozone-Safe or Safe for the Ozone Layer
    • Free-of a substance that causes environmental harm
Specifications

Specifications

Clear
Borosilicate Glass (Type 1)
Glass, Fused
Conical
100
Yes
12 mm
350 μL
Untreated
32 mm
9 mm
9 mm Screw Thread Caps - MicroLiter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions