missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mettler Toledo™ SevenGo™ Series Portable Meter: Model SG23
Characterized by user-friendliness and excellent ergonomics. Mettler Toledo SevenGo Series SG23 Portable Meter is watertight and robust instrument with high measurement accuracy.
Manufacturer: Mettler Toledo™ 51302601
Description
- Easy and intuitive operation
- pH range: 0 through +14.00
- Large, easy-to-read display screen
- Comprehensive range of practical accessories
- Modern GLP data management
- Revolutionary Intelligent Sensor Management (ISM™)
Specifications
±0.5% | |
±1mV | |
4 x AA | |
GLP/IP 67 | |
Determination of pH, mV, conductivity, TDS (total dissolved solids), salinity and specific resistance | |
pH: BNC/RCA, conductor: LTW 7-pin | |
Electrode Kit | |
0.01/1 | |
±0.5°C | |
0°C to 40°C | |
LCD | |
0.01 μS/cm to 500 mS/cm | |
0 to 80ppt | |
-1999 to 1999 mV | |
0.01 |
±0.5% | |
±0.01 | |
3 Points, 4 pre defined buffer group | |
99 GLP data records | |
Meter, batteries, wrist strap, test certificate and declaration of conformity, InLab Expert-Pro-ISM-IP67 and InLab 738-ISM (both IP67 and 1.8m fixed cable) | |
NTC | |
0.01/1 | |
1mV | |
30°F to 104°F | |
0.1°C | |
IP 67 | |
0 to 100megohm/cm | |
0.01mg/L to 300g/L | |
0 to 14 |
Promotions
-
Earn Up to $2,000 in Accessories with Purchase
Buy METTLER TOLEDO balances or analytical instruments, you’ll get up to $2,000 to spend on accessories.: 06/30/21
-
Earn Up to 5X the Fisher Scientific Buck$ with Purchase
Bundle your purchase of qualifying METTLER TOLEDO balances or analytical instruments to earn more.: 06/30/21
-
Earn Up to 5X the Fisher Scientific Buck$ with Purchase
Bundle your purchase of qualifying METTLER TOLEDO balances or analytical instruments to earn more.: 01/31/21