missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ Portable pH/Conductivity/Dissolved Oxygen/Ion Meters

Catalog No. p-3980700
Change view
Click to view available options
Model:
pH, ORP
S9 DO Meter
Seven2Go Pro S8
Seven2Go S2
Seven2Go S3
Seven2Go S4
SevenGo Duo SG23
SevenGo Duo SG68
SevenGo Duo SG78
SevenGo Duo SG98
Channels:
1
2
Product Type:
Conductivity Meter
DO Meter
DO Portable Meter
Meter with Biotech Kit
Meter with BOD Kit
Meter with Electrode Kit
Meter with Field Kit
Meter with Field Kit, 2m Cable
Meter with Field Kit, 5m Cable
Meter with Fluoride Kit
Meter with Food Kit
Meter with Light Kit
Meter with Standard Kit
pH/Conductivity Meter
pH/Ion/Cond Meter
pH/Ion/DO Meter
pH/Ion/DO/ORP Meter
pH/mV and Ion Meter
pH/mV Meter
29 product options available for selection
Product selection table with 29 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Channels Product Type
01917131 SevenGo Duo SG23 2 Meter with Electrode Kit
01917130 SevenGo Duo SG23 2 pH/Conductivity Meter
01915702 SevenGo Duo SG98 2 Meter with Field Kit, 2m Cable
01915703 SevenGo Duo SG98 2 Meter with Field Kit, 5m Cable
01917133 SevenGo Duo SG68 2 pH/Ion/DO Meter
01917134 SevenGo Duo SG68 2 Meter with Electrode Kit
01917137 SevenGo Duo SG78 2 Meter with Electrode Kit
01917138 SevenGo Duo SG78 2 Meter with Field Kit
01913970 Seven2Go S3 1 Conductivity Meter
01913971 Seven2Go S3 1 Meter with Standard Kit
01913974 Seven2Go S4 1 DO Meter
01913975 Seven2Go S4 1 Meter with Standard Kit
01913976 Seven2Go S4 1 Meter with Field Kit
01913987 S9 DO Meter 1 Meter with Standard Kit
01913988 S9 DO Meter 1 Meter with Field Kit
01913989 S9 DO Meter 1 Meter with BOD Kit
01913965 Seven2Go S2 1 pH/mV Meter
01913966 Seven2Go S2 1 Meter with Standard Kit
01913967 Seven2Go S2 1 Meter with Field Kit
01913968 Seven2Go S2 1 Meter with Food Kit
01913969 pH, ORP 1 Meter with Light Kit
01913981 Seven2Go Pro S8 1 pH/mV and Ion Meter
01913982 Seven2Go Pro S8 1 Meter with Standard Kit
01913983 Seven2Go Pro S8 1 Meter with Field Kit
01913984 Seven2Go Pro S8 1 Meter with Biotech Kit
01913985 Seven2Go Pro S8 1 Meter with Fluoride Kit
01914336 S9 DO Meter 1 DO Portable Meter
01917136 SevenGo Duo SG78 2 pH/Ion/Cond Meter
01915700 SevenGo Duo SG98 2 pH/Ion/DO/ORP Meter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 29 options available.
29 options
Catalog No. 01917131 Supplier METTLER TOLEDO™ Supplier No. 51302601

missing translation for 'validMainframeMsg'

Water and dust proof, IP67 rated meter for portable or field measurements. Portable meters compatible with pH, conductivity, dissolved oxygen, and ion selective electrodes.

  • Dust proof
  • Water proof
  • IP67 Rated
  • pH, Redox/ORP, conductivity, dissolved oxygen and ISE measuring capability (model dependent)
  • Compatible with a wide range of IP67-rated electrodes for any application
  • Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
  • Data storage available; capacity dependent on model
  • Export data to a printer or Excel file for Seven2Go Pro only
  • EasyDirect software available for data transfer (model dependent) for Seven2Go Pro only
  • Rubber holster available for single-handed measurements
  • uGo carrying and protecting case included in selective kits (see below for complete list); available for all portable meters

Specifications

Accuracy (Conductivity) ±0.5%
Accuracy (mV) ±1mV
Accuracy (pH) ±0.01
Accuracy (TDS) ±0.5%
Battery Type 4 x AA
Calibration Points 3 Points, 4 pre defined buffer group
Certifications/Compliance ISO 9001:2000
Data Management Details 99 GLP data records
Channels 2
Display Type LCD
IP Rating IP67
Measurement Modes ORP
Range (Conductivity) 0.01 μS/cm to 500 mS/cm
Range (mV) -1999 to 1999 mV
Range (pH) 0 to 14
Range (Resistivity) 0 to 100 Mohm/cm
Range (Salinity) 0 to 80 ppt
Range (TDS) 0.01 mg/L to 300 g/L
Resolution (pH) 0.01
Temperature Range (Metric) -5°C to +105°C
For Use With (Application) Determination of pH, mV, conductivity, TDS (total dissolved solids), salinity and specific resistance
Includes InLab Expert Pro ISM pH/temperature electrode; InLab 738-ISM conductivity/temperature electrode
Inputs pH: BNC/RCA, conductor: LTW 7-pin
Model SevenGo Duo SG23
Probe Type NTC
Product Type Meter with Electrode Kit
Resolution (Conductivity) 0.01/1
Resolution (mV) 1mV
Resolution (TDS) 0.01/1
Temperature Accuracy ±0.5°C
Temperature (English) Operating 30°F to 104°F
Temperature (Metric) Operating 0°C to 40°C
Temperature Resolution 0.1°C
Show More Show Less