Learn More
Brady™ Lockout Tagout Tags: NO HACER FUNCIONAR LOCKED OUT DO NOT ENERGIZE This lock/tag may only be removed by
GREENER_CHOICE
Heavy Duty Polyester (B-837) plastic tags are encapsulated in matte polyester laminate for superior durability during lockout and tagout situations.
$21.59 - $78.86
Specifications
Product Type Tag
Certifications/Compliance ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145; OSHA 1910.147
Quantity 25 Pk.
View More Specs
Products 3
Catalog Number Mfr. No. Material Price Quantity  
Catalog Number Mfr. No. Material Price Quantity  
19113216
View Documents
Brady™
BRD133562
Heavy-duty Polyester (B-837)
Pack of 25 Each for $78.86
Only null left
19-113-219
View Documents
Brady™
133565
Economy Polyester (B-851)
Pack of 25 Each for $60.30
Only null left
19-113-221
View Documents
Brady™
133567
Paper Cardstock
Pack of 25 Each for $21.59
Only null left
Description
Description
  • Write-on polyester laminate protects the graphics, wipes clean and withstands water, grease and extreme temperatures
  • Durable heavy duty polyester material with brass grommets exceeds OSHA's 50 lb pull force requirement for tagout devices
  • Excellent chemical, oil and water resistance
  • Service temperature of -40°F to 176°F (-40°C to 80°C)
Specifications
Specifications
Tag
ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145; OSHA 1910.147
25 Pk.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'