Learn More
Labconco™ G4 Replacement Prefilters
For use with Purifier™ Logic™ Biosafety Cabinet
Supplier: Labconco™ 3406202
Description
- Includes pleated disposable filter with Merv 8 rating
- Replacement filter cells for Purifier Logic™ and Cell Logic™ Disposable Prefilter Kit
Greener Choice Claims
- Environmental benefits include:
- Free-of a substance that causes environmental harm
- Non-Toxic
- Ozone-Safe or Safe for the Ozone Layer
- Recyclable
- Recycled Content
- Renewable Energy
- Renewable Materials
Specifications
G4 Prefilter | |
4 lb. | |
Pleated disposable filter with Merv 8 rating |
Purifier Logic Biosafety Cabinet (5 ft.) | |
1.8 kg | |
Replacement Prefilter |