Learn More
Labconcoâ„¢ G4 Replacement Prefilters
For use with Purifierâ„¢ Logicâ„¢ Biosafety Cabinet
$172.07 - $345.02
Specifications
Filter Type G4 Prefilter
Includes Pleated disposable filter with Merv 8 rating
Product Type Replacement Prefilter
View More Specs
Products 4
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
34-062-01
View Documents
Labconcoâ„¢
3406201
Purifier Logic Biosafety Cabinet (4 ft.)
Each for $227.59
Only null left
34-062-00
View Documents
Labconcoâ„¢
3406200
Purifier Logic Biosafety Cabinet (3 ft.)
Each for $172.07
Only null left
34-062-02
View Documents
Labconcoâ„¢
3406202
Purifier Logic Biosafety Cabinet (5 ft.)
Each for $281.80
Only null left
34-062-03
View Documents
Labconcoâ„¢
3406203
Purifier Logic Biosafety Cabinet (6 ft.)
Each for $345.02
Only null left
Description
Description
  • Includes pleated disposable filter with Merv 8 rating
  • Replacement filter cells for Purifier Logicâ„¢ and Cell Logicâ„¢ Disposable Prefilter Kit
Specifications
Specifications
G4 Prefilter
Pleated disposable filter with Merv 8 rating
Replacement Prefilter
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'