Learn More
Labconco™ G4 Replacement Prefilters
For use with Purifier™ Logic™ Biosafety Cabinet
Supplier: Labconco™ 3406201
Description
- Includes pleated disposable filter with Merv 8 rating
- Replacement filter cells for Purifier Logic™ and Cell Logic™ Disposable Prefilter Kit
Specifications
G4 Prefilter | |
3 lb. | |
Pleated disposable filter with Merv 8 rating |
Purifier Logic Biosafety Cabinet (4 ft.) | |
1.4 kg | |
Replacement Prefilter |