Learn More
MilliporeSigma™ Isopore™ Membrane Filters, 5 μm

Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
$120.00 - $534.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Color | White |
Filter Type | Screen Filter |
Material | Polycarbonate |
Pore Size | 5 μm |
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity | ||||||
TMTP01300
|
MilliporeSigma™
TMTP01300 |
13 mm |
Pack of 100 Each for $120.00
|
|
||||||
TMTP02500
|
MilliporeSigma™
TMTP02500 |
25 mm |
Pack of 100 Each for $124.00
|
|
||||||
TMTP04700
|
MilliporeSigma™
TMTP04700 |
47 mm |
Pack of 100 Each for $196.00
|
|
||||||
TMTP09030
|
MilliporeSigma™
TMTP09030 |
90 mm |
Pack of 30 Each for $294.22
|
|
||||||
TMTP14250
|
MilliporeSigma™
TMTP14250 |
142 mm |
Pack of 50 Each for $534.00
|
|
||||||
Description
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
The Isopore™ membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore™ membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore™ membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore™ membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety
Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays
Specifications
Not Autoclavable | |
Screen Filter | |
5 μm | |
Air Analysis; Cell Biology; Cell Culture; Bioassays; Cytology; Environmental Analysis; Epifluorescence. | |
Hydrophilic | |
Hydrophilic |
White | |
Polycarbonate | |
Non-sterile | |
50.0 L/min/cm2 | |
Filter Membranes |