missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HisPur™ Cobalt Resin

Gently purify His-tagged recombinant fusion proteins using cobalt (Co2+) immobilized metal chelate affinity (IMAC) beads, spin columns and plates.

Supplier:  Thermo Scientific™ 89964

 View more versions of this product

Catalog No. PI89964


Only null left
Explore available promotions

Description
Show More
Specifications

Specifications

Store at 4°C
HisPur™
Recombinant
Resin
Bottle
Liquid Suspension
10 mL
His-Tagged Protein
Metal Chelate, Divalent Cobalt
Affinity, Agarose Resin
Product Suggestions

Product Suggestions

Videos
SDS
Documents Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.