missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Newborn Calf Serum, New Zealand origin DFS Item

Gibco sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

$66.00 - $468.00

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Species Cattle/Bovine
Age Newborn (20 days or less)
Country of Origin New Zealand
Format Bottle
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
16010142 DFS Item
View Documents
Gibco™
16010142
1 L
Each for $468.00
Only null left
 
16010159 DFS Item
View Documents
Gibco™
16010159
500 mL
Each for $242.00
Only null left
 
16010167 DFS Item
View Documents
Gibco™
16010167
100 mL
Each for $66.00
Only null left
 
Description

Description

Gibco sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

  • Sera Category: Secure
  • Origin: New Zealand
  • Collected from calves 20 days old or less.

Order Info

Shipping Condition: Dry Ice

Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Newborn (20 days or less)
Bottle
Sterile
Cattle/Bovine
New Zealand
Standard (Sterile-filtered)
Frozen
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'