missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Newborn Calf Serum, New Zealand origin
GREENER_CHOICE

Catalog No. 16010167
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16010167 100 mL
16010142 1 L
16010159 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 16010167 Supplier Gibco™ Supplier No. 16010167

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gibco sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

  • Sera Category: Secure
  • Origin: New Zealand
  • Collected from calves 20 days old or less.

Order Info

Shipping Condition: Dry Ice

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Species Cattle/Bovine
Age Newborn (20 days or less)
Country of Origin New Zealand
Form Liquid
Format Bottle
Product Type Newborn Calf Bovine Serum
Quantity 100 mL
Serum Treatment Standard (Sterile-filtered)
Sterility Sterile
Shipping Condition Frozen
Show More Show Less
Will improper storage of my tissue culture reagents affect the growth rate of my culture?

Yes. Store animal sera at -5 to -20 degrees C. Store media at 2 to 8 degrees C; use within recommended shelf life period. Store complete media (supplemented) at 2 to 8 degrees C, and for complete medium the recommended shelf life is 2 to 4 weeks. Additionally, minimize exposure of sera and media to light.

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.