Learn More
Gibco™ Horse Serum, New Zealand origin

Description
Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
- Sera Category: Specialty
- Origin: New Zealand
- EIA tested (negative) serum from a donor herd
Order Info
Shipping Condition: Dry ice
Specifications
Specifications
| Content And Storage | Storage conditions: ≤-10°C, Shipping conditions: Frozen |
| Species | Horse/Equine, Horse |
| Age | Adult |
| Country of Origin | New Zealand |
| Form | Liquid |
| Product Type | Horse Serum |
| Quantity | 100 mL |
| Serum Treatment | Standard (Sterile-filtered) |
| Sterility | Sterile |
| Shipping Condition | Frozen |
Frequently Asked Questions (FAQs)
No. Our horse serum is collected from a controlled herd. Should one of the animals require treatment with an antibiotic, that animal is removed from the drawing program for the full witholding period of any drug administered.
Yes. Store animal sera at -5 to -20 degrees C. Store media at 2 to 8 degrees C; use within recommended shelf life period. Store complete media (supplemented) at 2 to 8 degrees C, and for complete medium the recommended shelf life is 2 to 4 weeks. Additionally, minimize exposure of sera and media to light.
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.