missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Horse Serum, New Zealand origin
GREENER_CHOICE

Catalog No. p-4921445
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16050130 100 mL
16050122 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16050130 Supplier Gibco™ Supplier No. 16050130
Only null left

Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support.

Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera Category: Specialty
  • Origin: New Zealand
  • EIA tested (negative) serum from a donor herd

Order Info

Shipping Condition: Dry ice

Specifications

Content And Storage Storage conditions: ≤-10°C, Shipping conditions: Frozen
Species Horse/Equine, Horse
Age Adult
Country of Origin New Zealand
Form Liquid
Product Type Horse Serum
Quantity 100 mL
Serum Treatment Standard (Sterile-filtered)
Sterility Sterile
Shipping Condition Frozen

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.