missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Horse Serum, New Zealand origin DFS Item
GREENER_CHOICE

Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support.

Supplier:  Gibco™ 16050122

 View more versions of this product

Catalog No. 16050122


Only null left
Explore available promotions

Description

Description

Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera Category: Specialty
  • Origin: New Zealand
  • EIA tested (negative) serum from a donor herd
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Specifications

Specifications

Storage conditions: ≤-10°C, Shipping conditions: Frozen
Adult
Liquid
500 mL
Sterile
Horse/Equine, Horse
New Zealand
Horse Serum
Standard (Sterile-filtered)
Frozen
Product Suggestions

Product Suggestions

Videos
SDS
Documents Promotions

Promotions

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.