missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DPBS, powder, no calcium, no magnesium DFS Item

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.

$32.01 - $126.00

Spécifications

Chemical Name or Material DPBS
For Use With (Application) Mammalian Cell Culture
pH 7.2 to 7.7
Physical Form Powder
Product Line Gibco™
voir les spécifications

Produits 3
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
21600010 DFS Item
Afficher les documents
Gibco™
21600010
10 x 1 L
chaque for $99.25
Il en reste null
 
21600069 DFS Item
Afficher les documents
Gibco™
21600069
10 L
chaque for $32.01
Il en reste null
 
21600044 DFS Item
Afficher les documents
Gibco™
21600044
50 L
chaque for $126.00
Il en reste null
 
Description

Description

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

This DPBS is modified as follows:

  • Without: Calcium, Magnesium, Phenol Red

The complete formulation is available.

Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).

Order Info

Shipping Condition: Room Temperature

Compliance

Manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Spécifications

Spécifications

DPBS
7.2 to 7.7
Gibco™
No Sodium Pyruvate
265 - 310 mOsm/kg
Mammalian Cell Culture
Powder
Dulbecco's Phosphate Buffered Saline
cGMP-compliant under the ISO 13485 standard
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.