missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gallium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$290.12 - $500.50
Chemical Identifiers
CAS | 7440-55-3 |
---|---|
Molecular Formula | Ga |
Molecular Weight (g/mol) | 69.72 |
MDL Number | MFCD00134045 |
InChI Key | GYHNNYVSQQEPJS-UHFFFAOYSA-N |
PubChem CID | 5360835 |
ChEBI | CHEBI:49631 |
IUPAC Name | gallium |
SMILES | [Ga] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
PLGA2-2Y
|
SPEX CertiPrep Group LLC
PLGA22Y |
125 mL | Triple-leached LDPE bottle |
Each for $290.12
|
|
|||||
PLGA2-2X
|
SPEX CertiPrep Group LLC
PLGA22X |
500 mL | Triple-leached LDPE bottle |
Each for $500.50
|
|
|||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ga | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7440-55-3 | |
69.72 | |
GYHNNYVSQQEPJS-UHFFFAOYSA-N | |
CHEBI:49631 | |
[Ga] |
Ga | |
MFCD00134045 | |
5360835 | |
gallium |
Specifications
Triple-leached LDPE bottle | |
Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
0.1,2.0,97.9 | |
Ga | |
AA, ICP | |
Certified Reference Material | |
[Ga] | |
gallium | |
125 mL | |
CHEBI:49631 | |
Matrix Concentration Cat. No. 2-5% HNO3 1000μg/mL PLGA2-2Y, -2X 10,000 PLGA2-3Y, -3X |
7440-55-3,7697-37-2,7732-18-5 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
0.1,2.0,97.9 | |
MFCD00134045 | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
GYHNNYVSQQEPJS-UHFFFAOYSA-N | |
2% HNO3 | |
69.72 | |
5360835 | |
Assurance | |
Gallium Standard |
Safety and Handling
Causes skin irritation, serious eye irritation
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only