missing translation for 'onlineSavingsMsg'
Learn More

Gallium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLGA22Y

 View more versions of this product

Catalog No. PLGA2-2Y


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ga1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Gallium Standard
7440-55-3,7697-37-2,7732-18-5
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
0.1,2.0,97.9
MFCD00134045
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
GYHNNYVSQQEPJS-UHFFFAOYSA-N
2% HNO3
69.72
5360835
Assurance
Triple-leached LDPE bottle
Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
0.1,2.0,97.9
Ga
AA, ICP
Certified Reference Material
[Ga]
gallium
125 mL
CHEBI:49631
Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLGA2-2Y, -2X    10,000    PLGA2-3Y, -3X

Chemical Identifiers

7440-55-3
69.72
GYHNNYVSQQEPJS-UHFFFAOYSA-N
CHEBI:49631
[Ga]
Ga
MFCD00134045
5360835
gallium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS Documents

Documents

Promotions

Promotions

Product intended for laboratory use only