missing translation for 'onlineSavingsMsg'
Learn More

Traceable™ ISO 17034 (formerly ISO Guide 34) Color-Coded CRM pH Buffers (Certified Reference Material), Fisherbrand™

Performs accurate, reproducible pH calibrations with color-coded bottles. Fisherbrand™ Traceable, Color-Coded buffers are Certified Reference Material pH Buffers.Compatible with all makes of pH instruments and electrodes.

$66.32 - $67.53

Specifications

For Use With (Application) Calibration
For Use With (Equipment) pH Instruments and Electrodes
Physical Form Liquid
Quantity 473 mL
Solution Type Buffer
View More Specs

Products 3
Catalog Number Mfr. No. pH Color Price Quantity  
Catalog Number Mfr. No. pH Color Price Quantity  
15-078-280
View Documents
Control Company
9876949
4.005 Red
Each for $66.32
Only null left
 
15-078-281
View Documents
Control Company
9876950
7.000 Yellow
Each for $66.32
Only null left
 
15-078-282
View Documents
Control Company
9876951
10.012 Blue
Each for $67.53
Only null left
 
Description

Description

Performs accurate, reproducible pH calibrations with color-coded bottles. Fisherbrand™ Traceable, Color-Coded buffers are Certified Reference Material pH Buffers. Compatible with all makes of pH instruments and electrodes.

  • Certified Reference Material (CRM) solutions meets all test requirements for federal, state and local agencies
  • 100% compatible with all makes of pH instruments and electrodes
  • Long term buffer stability and buffer resistance to temperature changes during pH meter calibration
  • Manufactured to IUPAC (International Union of Pure and Applied Chemistry) formulation
  • Triple-layered proprietary packaging (aluminum, foil, PET and low-density polyethylene bag) prevents contamination by permeable gases
  • Each 16 ounce bottle or six-pack single-use 100ml containers is supplied with individual temperature chart, traceability information and Traceable™ Certificate
Specifications

Specifications

Calibration
Liquid
Buffer
pH Instruments and Electrodes
473 mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'