missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Traceable™ ISO 17034 (formerly ISO Guide 34) Color-Coded CRM pH Buffers (Certified Reference Material), Fisherbrand™
Performs accurate, reproducible pH calibrations with color-coded bottles. Fisherbrand™ Traceable, Color-Coded buffers are Certified Reference Material pH Buffers.Compatible with all makes of pH instruments and electrodes.
Supplier: Control Company 9876949
Description
Performs accurate, reproducible pH calibrations with color-coded bottles. Fisherbrand™ Traceable, Color-Coded buffers are Certified Reference Material pH Buffers. Compatible with all makes of pH instruments and electrodes.
- Certified Reference Material (CRM) solutions
- Meets all test requirements for federal, state and local agencies
- 100% compatible with all makes of pH instruments and electrodes
- Long term buffer stability and buffer resistance to temperature changes during pH meter calibration
- Manufactured to IUPAC (International Union of Pure and Applied Chemistry) formulation
- Triple-layered proprietary packaging (aluminum, foil, PET and low-density polyethylene bag) prevents contamination by permeable gases
- Supplied with individual temperature chart, traceability information and Traceable™ Certificate
Specifications
Buffer Standards pH 4.005(CRM) | |
pH Instruments and Electrodes | |
Liquid | |
Buffer | |
'Certified and Traceable™ ' âISO 17034 (formerly ISO Guide 34)â and âCertified Reference Materialâ |
Red | |
4.005 | |
473 mL | |
473mL (16 oz.) bottle (ea) |