missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ferric Chloride CS, USP/EP Yellow Primary Solution, 1mL = 45.0mg FeCl3·6H2O, Ricca Chemical
Manufacturer: Ricca Chemical Company RI311032
Specifications
Ferric Chloride | |
10025-77-1,7647-01-0,7732-18-5 | |
Liquid | |
Amber Glass | |
1L |
Yellow Primary Solution, Colorimetric Solution | |
1 mL = 45.0mg FeCl3·6H2O | |
EP/USP | |
Amber |
Chemical Identifiers
10025-77-1 |