missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Chloride CS, USP/EP Yellow Primary Solution, Ricca Chemical
1 mL = 45.0 mg FeCl3.6H2O
$204.61 - $454.82
Chemical Identifiers
| CAS | 10025-77-1 |
|---|---|
| Molecular Formula | Cl3FeH12O6 |
| Molecular Weight (g/mol) | 270.29 |
| MDL Number | MFCD00149712 |
| InChI Key | NQXWGWZJXJUMQB-UHFFFAOYSA-K |
| IUPAC Name | iron(3+) hexahydrate trichloride |
| SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
3110-4
|
Ricca Chemical Company
31104 |
120 mL | Amber Glass Bottle |
N/A
|
|
||||
|
3110-16
|
Ricca Chemical Company
311016 |
500 mL | Amber Glass Bottle |
N/A
|
|
||||
|
3110-32
|
Ricca Chemical Company
311032 |
1 L | Amber Glass Bottle |
N/A
|
|
||||
Chemical Identifiers
| 10025-77-1 | |
| 270.29 | |
| NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
| Cl3FeH12O6 | |
| MFCD00149712 | |
| iron(3+) hexahydrate trichloride |
Specifications
| Amber | |
| Cl3FeH12O6 | |
| NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
| iron(3+) hexahydrate trichloride | |
| 1 mL = 45.0 mg FeCl3·6H2O | |
| Amber Glass Bottle | |
| 120 mL | |
| Ferric Chloride, Yellow Primary Solution, Colorimetric Solution |
| 10025-77-1 , 7647-01-0 , 7732-18-5 | |
| MFCD00149712 | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] | |
| 270.29 | |
| EP/USP/Colorimetric Solution | |
| Acidic | |
| Liquid |